Assay Details
| Gene Symbol: PPP1R3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_423102.5 | XP_423102.2 | 2 | 1258 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R3C| Gene Symbol: PPP1R3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_423102.5 | XP_423102.2 | 2 | 1258 | 107 | |
Set Membership: |
|