Assay Details
| Gene Symbol: LSM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417768.5 | XP_417768.1 | 1 - 2 | 36 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_417768.5 transcripts.
Set Membership: |
|

LSM10
Genomic Map