Assay Details
| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149299.1 | XP_025005067.1 | 8 - 9 | 1063 | 98 | |
| XM_419525.6 | XP_419525.2 | 9 - 10 | 1174 | 98 | ||
More Information
Set Membership: |
|

NDUFAF7
Genomic Map