Assay Details
| Gene Symbol: LOC101751456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143530.1 | XP_024999298.1 | 6 - 7 | 658 | 79 | |
| Gene Symbol: CCT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298317.2 | XP_015153803.2 | 6 - 7 | 502 | 79 | |
More Information
Set Membership: |
|

LOC101751456
Genomic Map