Assay Details
| Gene Symbol: C8orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282850.2 | XP_015138336.1 | 1 - 2 | 350 | 108 | |
| XM_015282852.2 | XP_015138338.1 | 1 - 2 | 353 | 108 | ||
More Information
Set Membership: |
|

C8orf59
Genomic Map