Assay Details
| Gene Symbol: DAPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299832.2 | XP_015155318.2 | 9 | 2403 | 100 | |
| XM_025144345.1 | XP_025000113.1 | 9 | 2382 | 100 | ||
More Information
Set Membership: |
|

DAPK3
Genomic Map