Assay Details
| Gene Symbol: NT5C1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947880.3 | XP_004947937.1 | 6 | 4156 | 88 | |
| XM_015297909.2 | XP_015153395.1 | 6 | 2817 | 88 | ||
| XM_025143111.1 | XP_024998879.1 | 6 | 2898 | 88 | ||
| XM_417822.6 | XP_417822.3 | 6 | 2978 | 88 | ||
More Information
Set Membership: |
|

NT5C1A
Genomic Map