Assay Details
| Gene Symbol: WASF3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278585.2 | XP_015134071.2 | 8 - 9 | 1509 | 93 | |
| XM_015278586.2 | XP_015134072.2 | 8 - 9 | 1450 | 93 | ||
More Information
Set Membership: |
|

WASF3L
Genomic Map