Assay Details
| Gene Symbol: AOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004948624.3 | XP_004948681.1 | 4 - 5 | 3185 | 87 | |
| XM_015299613.2 | XP_015155099.1 | 4 - 5 | 3185 | 87 | ||
More Information
Set Membership: |
|

AOC3
Genomic Map