Assay Details
| Gene Symbol: CLAPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274310.2 | XP_015129796.2 | 2 - 3 | 513 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLAPS2| Gene Symbol: CLAPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274310.2 | XP_015129796.2 | 2 - 3 | 513 | 66 | |
Set Membership: |
|