Assay Details
| Gene Symbol: PCBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292855.2 | XP_015148341.1 | 13 - 14 | 2513 | 62 | |
| XM_015292856.2 | XP_015148342.1 | 13 - 14 | 2493 | 62 | ||
| XM_025154689.1 | XP_025010457.1 | 13 - 14 | 2513 | 62 | ||
| XM_025154691.1 | XP_025010459.1 | 12 - 13 | 2406 | 62 | ||
More Information
Set Membership: |
|

PCBP4
Genomic Map