Assay Details
| Gene Symbol: LOC421195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419273.5 | XP_419273.3 | 41 - 42 | 7187 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC421195| Gene Symbol: LOC421195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419273.5 | XP_419273.3 | 41 - 42 | 7187 | 119 | |
Set Membership: |
|