Assay Details
| Gene Symbol: ANGPT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235002.5 | XP_001235003.2 | 9 | 2541 | 133 | |
| XM_025142329.1 | XP_024998097.1 | 9 | 2520 | 133 | ||
More Information
Set Membership: |
|

ANGPT4
Genomic Map