Assay Details
| Gene Symbol: CA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233319.5 | XP_001233320.2 | 10 - 11 | 1088 | 55 | |
| XM_004937157.3 | XP_004937214.1 | 10 - 11 | 1091 | 55 | ||
More Information
Set Membership: |
|

CA9
Genomic Map