Assay Details
| Gene Symbol: MAPKAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299062.2 | XP_015154548.1 | 12337 | 79 | ||
| XM_025143775.1 | XP_024999543.1 | 1982 | 79 | |||
More Information
Set Membership: |
|

MAPKAPK2
Genomic Map