Assay Details
| Gene Symbol: HIST1H2BF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232984.4 | XP_001232985.1 | 1 | 517 | 130 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2BF| Gene Symbol: HIST1H2BF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232984.4 | XP_001232985.1 | 1 | 517 | 130 | |
Set Membership: |
|