Assay Details
| Gene Symbol: ADAMTSL2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943219.3 | XP_004943276.3 | 16 - 17 | 3058 | 101 | |
| XM_025153225.1 | XP_025008993.1 | 17 - 18 | 6403 | 101 | ||
| XM_025153227.1 | XP_025008995.1 | 17 - 18 | 6398 | 101 | ||
| XM_025153228.1 | XP_025008996.1 | 15 - 16 | 2749 | 101 | ||
| XM_422263.6 | XP_422263.6 | 16 - 17 | 3055 | 101 | ||
More Information
Set Membership: |
|

ADAMTSL2L
Genomic Map