Assay Details
| Gene Symbol: PFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285044.2 | XP_015140530.1 | 4 - 5 | 432 | 102 | |
| XM_025149107.1 | XP_025004875.1 | 4 - 5 | 475 | 102 | ||
More Information
Set Membership: |
|

PFN4
Genomic Map