Assay Details
| Gene Symbol: LOC101747868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940078.3 | XP_004940135.1 | 3 | 1833 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101747868| Gene Symbol: LOC101747868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940078.3 | XP_004940135.1 | 3 | 1833 | 113 | |
Set Membership: |
|