Assay Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280899.2 | XP_015136385.1 | 5 - 6 | 804 | 77 | |
| XM_417284.6 | XP_417284.2 | 5 - 6 | 514 | 77 | ||
More Information
Set Membership: |
|

RNF175
Genomic Map