Assay Details
| Gene Symbol: C2H8ORF84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_429743.6 | XP_429743.4 | 1 - 2 | 524 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2H8ORF84| Gene Symbol: C2H8ORF84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_429743.6 | XP_429743.4 | 1 - 2 | 524 | 102 | |
Set Membership: |
|