Assay Details
Gene Symbol: SLC35G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942154.3 | XP_004942211.2 | 3 | 4670 | 87 | |
XM_004942155.3 | XP_004942212.2 | 3 | 4547 | 87 | ||
XM_015288857.2 | XP_015144343.1 | 3 | 4679 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
