Assay Details
| Gene Symbol: TBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155713.1 | XP_025011481.1 | 8 | 4892 | 118 | |
| XM_025155714.1 | XP_025011482.1 | 11 | 4815 | 118 | ||
| XM_025155715.1 | XP_025011483.1 | 10 | 4621 | 118 | ||
| XM_025155716.1 | XP_025011484.1 | 8 | 4841 | 118 | ||
More Information
Set Membership: |
|

TBX1
Genomic Map