Assay Details
| Gene Symbol: TTLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935527.3 | XP_004935584.1 | 3 | 1908 | 96 | |
| XM_004935528.3 | XP_004935585.1 | 3 | 1225 | 96 | ||
| XM_004935530.3 | XP_004935587.1 | 3 | 1180 | 96 | ||
More Information
Set Membership: |
|

TTLL2
Genomic Map