Assay Details
| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298030.2 | XP_015153516.1 | 11 | 4208 | 94 | |
| XM_015298039.2 | XP_015153525.1 | 11 | 3832 | 94 | ||
More Information
Set Membership: |
|

GABRR3
Genomic Map