Assay Details
| Gene Symbol: MSMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942118.3 | XP_004942175.2 | 3 - 4 | 283 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSMB| Gene Symbol: MSMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942118.3 | XP_004942175.2 | 3 - 4 | 283 | 116 | |
Set Membership: |
|