Assay Details
| Gene Symbol: S100A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004948219.3 | XP_004948276.1 | 1 - 2 | 875 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004948219.3 transcripts.
Set Membership: |
|

S100A4
Genomic Map