Assay Details
| Gene Symbol: RTN4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415292.6 | XP_415292.4 | 2 | 2123 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTN4R| Gene Symbol: RTN4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415292.6 | XP_415292.4 | 2 | 2123 | 101 | |
Set Membership: |
|