Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273267.2 | XP_015128753.1 | 5 | 1145 | 67 | |
| XM_025146118.1 | XP_025001886.1 | 5 | 1102 | 67 | ||
More Information
Set Membership: |
|

THAP8
Genomic Map