Assay Details
| Gene Symbol: LOC101747820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150444.1 | XP_025006212.1 | 2 - 3 | 1610 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101747820| Gene Symbol: LOC101747820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150444.1 | XP_025006212.1 | 2 - 3 | 1610 | 62 | |
Set Membership: |
|