Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292653.2 | XP_015148139.1 | 7 - 8 | 995 | 97 | |
| XM_015292654.2 | XP_015148140.1 | 6 - 7 | 907 | 97 | ||
| XM_015292655.2 | XP_015148141.1 | 5 - 6 | 574 | 97 | ||
| XM_015292656.2 | XP_015148142.1 | 5 - 6 | 444 | 97 | ||
More Information
Set Membership: |
|

FBXO31
Genomic Map