Assay Details
| Gene Symbol: OTUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001236781.5 | XP_001236782.2 | 6 - 7 | 653 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTUB1| Gene Symbol: OTUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001236781.5 | XP_001236782.2 | 6 - 7 | 653 | 60 | |
Set Membership: |
|