Assay Details
| Gene Symbol: PTCHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425978.6 | XP_425978.4 | 4 | 4203 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTCHD3| Gene Symbol: PTCHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425978.6 | XP_425978.4 | 4 | 4203 | 111 | |
Set Membership: |
|