Assay Details
| Gene Symbol: RAB37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642366.3 | XP_003642414.1 | 9 | 1493 | 130 | |
| XM_004946346.3 | XP_004946403.2 | 10 | 2877 | 130 | ||
| XM_015295475.2 | XP_015150961.1 | 11 | 1775 | 130 | ||
| XM_025141859.1 | XP_024997627.1 | 11 | 5126 | 130 | ||
| XM_025141860.1 | XP_024997628.1 | 12 | 5059 | 130 | ||
| XM_025141861.1 | XP_024997629.1 | 10 | 5018 | 130 | ||
| XM_025141862.1 | XP_024997630.1 | 10 | 5060 | 130 | ||
| XM_025141863.1 | XP_024997631.1 | 10 | 4950 | 130 | ||
| XM_025141864.1 | XP_024997632.1 | 11 | 3900 | 130 | ||
| XM_025141865.1 | XP_024997633.1 | 9 | 4837 | 130 | ||
| XM_025141867.1 | XP_024997635.1 | 10 | 1829 | 130 | ||
| XM_025141868.1 | XP_024997636.1 | 9 | 2706 | 130 | ||
| XM_025141869.1 | XP_024997637.1 | 10 | 3724 | 130 | ||
More Information
Set Membership: |
|

RAB37
Genomic Map