Assay Details
| Gene Symbol: SLC6A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293329.2 | XP_015148815.1 | 11 - 12 | 1716 | 60 | |
| GenBank mRNA | JN628439.2 | - | 10 - 11 | 1432 | 60 | |
More Information
Set Membership: |
|

SLC6A8
Genomic Map