Assay Details
Gene Symbol: SRXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015296474.2 | XP_015151960.1 | 2 | 1217 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SRXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015296474.2 | XP_015151960.1 | 2 | 1217 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |