Assay Details
| Gene Symbol: MAP6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277169.2 | XP_015132655.1 | 2 - 3 | 1051 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP6D1| Gene Symbol: MAP6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277169.2 | XP_015132655.1 | 2 - 3 | 1051 | 121 | |
Set Membership: |
|