Assay Details
| Gene Symbol: CHST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287146.2 | XP_015142632.1 | 2 | 1866 | 100 | |
| XM_015287147.2 | XP_015142633.1 | 2 | 1917 | 100 | ||
More Information
Set Membership: |
|

CHST1
Genomic Map