Assay Details
| Gene Symbol: ADAMTS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298130.2 | XP_015153616.1 | 9 | 9320 | 162 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADAMTS8| Gene Symbol: ADAMTS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298130.2 | XP_015153616.1 | 9 | 9320 | 162 | |
Set Membership: |
|