Assay Details
| Gene Symbol: CRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290407.1 | XP_015145893.1 | 6 - 7 | 613 | 75 | |
| XM_015290408.2 | XP_015145894.2 | 6 - 7 | 573 | 75 | ||
| GenBank mRNA | BX931646.1 | - | 5 - 6 | 723 | 75 | |
More Information
Set Membership: |
|

CRIP2
Genomic Map