Assay Details
| Gene Symbol: MMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414787.5 | XP_414787.2 | 2 - 3 | 401 | 55 | |
| GenBank mRNA | BX931694.1 | - | 2 - 3 | 324 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MMD2| Gene Symbol: MMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414787.5 | XP_414787.2 | 2 - 3 | 401 | 55 | |
| GenBank mRNA | BX931694.1 | - | 2 - 3 | 324 | 55 | |
Set Membership: |
|