Assay Details
| Gene Symbol: CYTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152505.1 | XP_025008273.1 | 7 - 8 | 971 | 69 | |
| XM_426609.5 | XP_426609.3 | 9 - 10 | 4279 | 69 | ||
| GenBank mRNA | BX931951.1 | - | 5 - 6 | 701 | 69 | |
More Information
Set Membership: |
|

CYTIP
Genomic Map