Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942691.3 | XP_004942748.2 | 2 - 3 | 418 | 72 | |
| XM_004942692.3 | XP_004942749.1 | 2 - 3 | 392 | 72 | ||
| XM_025152688.1 | XP_025008456.1 | 2 - 3 | 418 | 72 | ||
| GenBank mRNA | BX932139.1 | - | 2 - 3 | 411 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025152688.1 transcripts.
Set Membership: |
|

ZNF385B
Genomic Map