Assay Details
| Gene Symbol: NAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015272486.2 | XP_015127972.2 | 4 - 5 | 378 | 54 | |
| GenBank mRNA | BX932246.2 | - | 4 - 5 | 367 | 54 | |
| BX935921.1 | - | 4 - 5 | 367 | 54 | ||
More Information
Set Membership: |
|

NAPA
Genomic Map