Assay Details
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947704.3 | XP_004947761.1 | 4 - 5 | 800 | 59 | |
XM_025143139.1 | XP_024998907.1 | 4 - 5 | 784 | 59 | ||
GenBank mRNA | BX932283.1 | - | 3 - 4 | 390 | 59 | |
BX934537.2 | - | 3 - 4 | 748 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
