Assay Details
| Gene Symbol: MYCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232873.5 | XP_001232874.2 | 3 - 4 | 230 | 104 | |
| GenBank mRNA | BX932400.1 | - | 2 - 3 | 269 | 104 | |
| BX933751.2 | - | 3 - 4 | 163 | 104 | ||
| BX950775.1 | - | 2 - 3 | 138 | 104 | ||
More Information
Set Membership: |
|

MYCBP
Genomic Map