Assay Details
| Gene Symbol: MIF4GD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295451.2 | XP_015150937.1 | 5 - 6 | 599 | 63 | |
| XM_015295452.2 | XP_015150938.1 | 5 - 6 | 1090 | 63 | ||
| XM_015295453.2 | XP_015150939.1 | 5 - 6 | 638 | 63 | ||
| XM_015295454.2 | XP_015150940.1 | 5 - 6 | 1149 | 63 | ||
| GenBank mRNA | BX932459.2 | - | 5 - 6 | 589 | 63 | |
| BX936163.1 | - | 5 - 6 | 586 | 63 | ||
More Information
Set Membership: |
|

MIF4GD
Genomic Map