Assay Details
| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280936.2 | XP_015136422.1 | 7 - 8 | 938 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPX| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280936.2 | XP_015136422.1 | 7 - 8 | 938 | 56 | |
Set Membership: |
|