Assay Details
| Gene Symbol: TCP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143871.1 | XP_024999639.1 | 569 | 85 | ||
| XM_025143872.1 | XP_024999640.1 | 569 | 85 | |||
| XM_025143873.1 | XP_024999641.1 | 569 | 85 | |||
| XM_025143874.1 | XP_024999642.1 | 376 | 85 | |||
| XM_418022.6 | XP_418022.3 | 420 | 85 | |||
More Information
Set Membership: |
|

TCP11
Genomic Map