Assay Details
| Gene Symbol: LOC100857858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300314.2 | XP_015155800.2 | 1 - 2 | 50 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015300314.2 transcripts.
Set Membership: |
|

LOC100857858
Genomic Map